Learn More
Biomatik Corporation Human LTF (Lactoferrin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57150
Encompass_Preferred
The Human LTF (Lactoferrin) ELISA Kit allows for quantitative determination of LTF in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.313-20 ng/mL. Sensitivity: 0.188 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P02788. Restriction: For Research Use Only.
Catalog No. 50-150-0796