Learn More
Biomatik Corporation Human LH (Luteinizing Hormone) ELISA Kit, 96 tests
Catalog No. 501500797
Encompass_Preferred
The Human LH (Luteinizing Hormone) ELISA Kit allows for quantitative determination of LH in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.938-60m IU/mL. Sensitivity: 0.563m IU/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.
Catalog No. 50-150-0797 Supplier Biomatik Corporation Supplier No. EKF57151
The Human LH (Luteinizing Hormone) ELISA Kit allows for quantitative determination of LH in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.938-60m IU/mL. Sensitivity: 0.563m IU/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.