Learn More
Biomatik Corporation Human sRAGE (Solube Receptor for Advanced Glycation End product) ELISA Kit, 96 tests
Catalog No. 501500800
Encompass_Preferred
Human sRAGE (soluble Receptor for Advanced Glycation End product) ELISA Kit for detecting sRAGE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q15109. Target Synonyms: sRAGE; soluble Receptor for Advanced Glycation End product; RAGE; AGER; Advanced glycosylation end product-specific receptor; Receptor for advanced glycosylation end products. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0800 Supplier Biomatik Corporation Supplier No. EKF57154
Human sRAGE (soluble Receptor for Advanced Glycation End product) ELISA Kit for detecting sRAGE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q15109. Target Synonyms: sRAGE; soluble Receptor for Advanced Glycation End product; RAGE; AGER; Advanced glycosylation end product-specific receptor; Receptor for advanced glycosylation end products. Restrictions: For Research Use Only. Not for use in diagnostic procedures.