Learn More
Biomatik Corporation Human GFAP (Glial fibrillary acidic protein) ELISA Kit, 96 tests
Catalog No. 501500801
Encompass_Preferred
The Human GFAP (Glial fibrillary acidic protein) ELISA Kit allows for quantitative determination of GFAP in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.313-20 ng/mL. Sensitivity: 0.188 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P14136. Restriction: For Research Use Only.
Catalog No. 50-150-0801 Supplier Biomatik Corporation Supplier No. EKF57155
The Human GFAP (Glial fibrillary acidic protein) ELISA Kit allows for quantitative determination of GFAP in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.313-20 ng/mL. Sensitivity: 0.188 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P14136. Restriction: For Research Use Only.