Learn More
Biomatik Corporation Human IgA (Immunoglobulin A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57157
Encompass_Preferred
The Human IgA (Immunoglobulin A) ELISA Kit allows for quantitative determination of IgA in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.781-50 ng/mL. Sensitivity: 0.469 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P11912. Restriction: For Research Use Only.
Catalog No. 50-150-0803