Learn More
Biomatik Corporation Human cTnT/TNNT2 (Troponin T Type 2, Cardiac) ELISA Kit, 96 tests
Catalog No. 501500810
Encompass_Preferred
The Human cTnT/TNNT2 (Troponin T Type 2, Cardiac) ELISA Kit allows for quantitative determination of cTnT/TNNT2 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P45379. Restriction: For Research Use Only.
Catalog No. 50-150-0810 Supplier Biomatik Corporation Supplier No. EKF57164
The Human cTnT/TNNT2 (Troponin T Type 2, Cardiac) ELISA Kit allows for quantitative determination of cTnT/TNNT2 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P45379. Restriction: For Research Use Only.