Learn More
Biomatik Corporation Human IFN-β (Interferon beta) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57167
Encompass_Preferred
The Human IFN-β (Interferon beta) ELISA Kit allows for quantitative determination of IFN-β in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 62.5-4000 pg/mL. Sensitivity: 37.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P01574. Restriction: For Research Use Only.
Catalog No. 50-150-0813