Learn More
Biomatik Corporation Human FGF2 (Heparin-binding growth factor 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57173
Encompass_Preferred
The Human FGF2 (Heparin-binding growth factor 2) ELISA Kit allows for quantitative determination of FGF2 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 12.5-800 pg/mL. Sensitivity: 7.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P09038. Restriction: For Research Use Only.
Catalog No. 50-150-0819