Learn More
Biomatik Corporation Human CASP3 (Caspase-3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57175
Encompass_Preferred
The Human CASP3 (Caspase-3) ELISA Kit allows for quantitative determination of CASP3 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.313-20 ng/mL. Sensitivity: 0.188 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P42574. Restriction: For Research Use Only.
Catalog No. 50-150-0821