Learn More
Biomatik Corporation Human TNNI1 (Troponin I, slow skeletal muscle) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57186
Encompass_Preferred
Human TNNI1 (Troponin I, slow skeletal muscle) ELISA Kit for detecting TNNI1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 46.875-3000pg/ml. Sensitivity: 28.125pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P19237. Target Synonyms: TNNI1; Troponin I Type 1; Slow Skeletal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0832