Learn More
Biomatik Corporation Human TNFRSF8 (Tumor necrosis factor receptor superfamily member 8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57190
Encompass_Preferred
Human TNFRSF8 (Tumor necrosis factor receptor superfamily member 8) ELISA Kit for detecting TNFRSF8 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P28908. Target Synonyms: TNFRSF8; CD30; CD30 antigen; KI-1; CD30L receptor; cytokine receptor CD30; Ki-1 antigen; Lymphocyte activation antigen CD30; tumor necrosis factor receptor superfamily member 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0836