Learn More
Biomatik Corporation Human C3 (Complement C3) ELISA Kit, 96 tests
Catalog No. 501500846
Encompass_Preferred
The Human C3 (Complement C3) ELISA Kit allows for quantitative determination of C3 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 7.813-500 ng/mL. Sensitivity: 4.688 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P01024. Restriction: For Research Use Only.
Catalog No. 50-150-0846 Supplier Biomatik Corporation Supplier No. EKF57200
The Human C3 (Complement C3) ELISA Kit allows for quantitative determination of C3 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 7.813-500 ng/mL. Sensitivity: 4.688 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P01024. Restriction: For Research Use Only.