Learn More
Biomatik Corporation Human GUSβ (Beta-glucuronidase) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKF57223
The Human GUSβ (Beta-glucuronidase) ELISA Kit allows for quantitative determination of GUSβ in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.094 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P08236. Restriction: For Research Use Only.