Learn More
Biomatik Corporation Human GPX1 (Glutathione peroxidase 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57238
Encompass_Preferred
The Human GPX1 (Glutathione peroxidase 1) ELISA Kit allows for quantitative determination of GPX1 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.188 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P07203. Restriction: For Research Use Only.
Catalog No. 50-150-0849