Learn More
Biomatik Corporation Human CPB2 (Carboxypeptidase B2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57270
Encompass_Preferred
The Human CPB2 (Carboxypeptidase B2) ELISA Kit allows for quantitative determination of CPB2 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 3.12-200 ng/mL. Sensitivity: 1.875 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: Q96IY4. Restriction: For Research Use Only.
Catalog No. 50-150-0853