Learn More
Biomatik Corporation Human CTXII (Cross Linked C-telopeptide of Type II Collagen) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57279
Encompass_Preferred
The Human CTXII (Cross Linked C-telopeptide of Type II Collagen) ELISA Kit allows for quantitative determination of CTXII in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.094 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.
Catalog No. 50-150-0854