Learn More
Biomatik Corporation Human DNASE1 (Deoxyribonuclease-1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57315
Encompass_Preferred
The Human DNASE1 (Deoxyribonuclease-1) ELISA Kit allows for quantitative determination of DNASE1 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.094 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P24855. Restriction: For Research Use Only.
Catalog No. 50-150-0861