Learn More
Biomatik Corporation Human SFRP1 (Secreted frizzled-related protein 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57317
Encompass_Preferred
Human SFRP1 (Secreted frizzled-related protein 1) ELISA Kit for detecting SFRP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8N474. Target Synonyms: SFRP1; Secreted frizzled-related protein 1; Secreted apoptosis-related protein 2; FRP-1; sFRP-1; FRP; FRP1; FrzA; FRP-1; SARP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0863