Learn More
Biomatik Corporation Human LBP (Lipopolysaccharide-binding protein) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57347
Encompass_Preferred
The Human LBP (Lipopolysaccharide-binding protein) ELISA Kit allows for quantitative determination of LBP in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 3.125-200 ng/mL. Sensitivity: 1.875 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P18428. Restriction: For Research Use Only.
Catalog No. 50-150-0866