Learn More
Biomatik Corporation Human AMY1 (Alpha-amylase 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57364
Encompass_Preferred
The Human AMY1 (Alpha-amylase 1) ELISA Kit allows for quantitative determination of AMY1 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 1.563-100 ng/mL. Sensitivity: 0.938 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.
Catalog No. 50-150-0867