Learn More
Biomatik Corporation Human FAM132B (Protein FAM132B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57370
Encompass_Preferred
The Human FAM132B (Protein FAM132B) ELISA Kit allows for quantitative determination of FAM132B in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.094 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: Q4G0M1. Restriction: For Research Use Only.
Catalog No. 50-150-0868