Learn More
Biomatik Corporation Human MTOR (Serine/threonine-protein kinase mTOR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57371
Encompass_Preferred
The Human MTOR (Serine/threonine-protein kinase mTOR) ELISA Kit allows for quantitative determination of MTOR in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.094 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P42345. Restriction: For Research Use Only.
Catalog No. 50-150-0869