Learn More
Biomatik Corporation Human RNASE3/ECP (Ribonuclease A3/Eosinophil Cationic Protein) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57395
Encompass_Preferred
Human RNASE3/ECP (Ribonuclease A3/Eosinophil Cationic Protein) ELISA Kit for detecting RNASE3; ECP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 39.063-2500pg/ml. Sensitivity: 23.438pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12724. Target Synonyms: RNASE3; ECP; Ribonuclease A3; Eosinophil Cationic Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0870