Learn More
Biomatik Corporation Human PRKCE (Protein kinase C epsilon type) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57496
Encompass_Preferred
The Human PRKCE (Protein kinase C epsilon type) ELISA Kit allows for quantitative determination of PRKCE in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100 ng/mL. Sensitivity: 0.938 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: Q02156. Restriction: For Research Use Only.
Catalog No. 50-150-0876