Learn More
Biomatik Corporation Human C3a (Complement Component 3a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57555
Encompass_Preferred
The Human C3a (Complement Component 3a) ELISA Kit allows for quantitative determination of C3a in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.313-20 ng/mL. Sensitivity: 0.188 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: Q16581. Restriction: For Research Use Only.
Catalog No. 50-150-0881