Learn More
Biomatik Corporation Human CYPA (Cyclophilin A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57586
Encompass_Preferred
The Human CYPA (Cyclophilin A) ELISA Kit allows for quantitative determination of CYPA in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 1.25-80 ng/mL. Sensitivity: 0.75 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P62937. Restriction: For Research Use Only.
Catalog No. 50-150-0884