Learn More
Biomatik Corporation Human IL-13 (Interleukin 13) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57642
Encompass_Preferred
The Human IL-13 (Interleukin 13) ELISA Kit allows for quantitative determination of IL-13 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P35225. Restriction: For Research Use Only.
Catalog No. 50-150-0892