Learn More
Biomatik Corporation Human Neopterin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57660
Encompass_Preferred
The Human Neopterin ELISA Kit allows for quantitative determination of Neopterin in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.094 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-0898