Learn More
Biomatik Corporation Human Pro-ADM (Proadrenomedullin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57697
Encompass_Preferred
The Human Pro-ADM (Proadrenomedullin) ELISA Kit allows for quantitative determination of Pro-ADM in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.781-50pmol/L. Sensitivity: 0.469pmol/L. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.
Catalog No. 50-150-0902