Learn More
Biomatik Corporation Human PTD (Pentosidine) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57699
Encompass_Preferred
The Human PTD (Pentosidine) ELISA Kit allows for quantitative determination of PTD in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 6.25-400 ng/mL. Sensitivity: 3.75 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-0903