Learn More
Biomatik Corporation Human TRACP-5b (Tartrate-resistant Acid Phosphatase 5b) ELISA Kit, 96 tests
Catalog No. 501500904
Encompass_Preferred
Human TRACP-5b (TartRate-resistant Acid Phosphatase 5b) ELISA Kit for detecting TRACP-5b in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20miu/ml. Sensitivity: 0.188miu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13686. Target Synonyms: MGC117378; TRAP; TRatPase; tartRate resistant acid phosphatase 5; tartRate-resistant acid ATPase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0904 Supplier Biomatik Corporation Supplier No. EKF57706
Human TRACP-5b (TartRate-resistant Acid Phosphatase 5b) ELISA Kit for detecting TRACP-5b in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20miu/ml. Sensitivity: 0.188miu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13686. Target Synonyms: MGC117378; TRAP; TRatPase; tartRate resistant acid phosphatase 5; tartRate-resistant acid ATPase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.