Learn More
Biomatik Corporation Human suPAR (soluble urokinase-type plasminogen activator receptor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57738
Encompass_Preferred
Human suPAR (soluble urokinase-type plasminogen activator receptor) ELISA Kit for detecting suPAR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q03405. Target Synonyms: soluble urokinase-type plasminogen activator receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0910