Learn More
Biomatik Corporation Human calprotectin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57741
Encompass_Preferred
The Human calprotectin ELISA Kit allows for quantitative determination of calprotectin in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 15.63-1000 ng/mL. Sensitivity: 9.375 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. Restriction: For Research Use Only.
Catalog No. 50-150-0913