Learn More
Biomatik Corporation Mouse Ang1 (Angiopoietin-1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57748
Encompass_Preferred
Mouse Ang1 (Angiopoietin-1) ELISA Kit for detecting Ang1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O08538. Target Synonyms: ANG1; Angiopoietin 1; ANG-1; ANGPT1; AGP1; AGPT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0920