Learn More
Biomatik Corporation Mouse CD40L (Cluster of Differentiation 40 Ligand) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57751
Encompass_Preferred
Mouse CD40L (Cluster of Differentiation 40 Ligand) ELISA Kit for detecting CD40L in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P27548. Target Synonyms: CD40L; TNFSF5; TNFSF5; CD40LG; CD154; HIGM1; IGM; IMD3; T-BAM; TRAP; Gp39. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0923