Learn More
Biomatik Corporation Mouse EPO (Erythropoietin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57753
Encompass_Preferred
Mouse EPO (Erythropoietin) ELISA Kit for detecting EPO in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07321. Target Synonyms: EPO; Erythropoietin; EP; MVCD2; Epoetin; Hematopoietin; Hemopoietin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0925