Learn More
Biomatik Corporation Mouse GDNF (Glial Cell Line Derived Neurotrophic Factor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57757
Encompass_Preferred
Mouse GDNF (Glial Cell Line Derived Neurotrophic Factor) ELISA Kit for detecting GDNF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.688-300pg/ml. Sensitivity: 2.813pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48540. Target Synonyms: GDNF; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; glial cell line-derived neurotrophic factor; HFB1-GDNF; hGDNF; HSCR3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0929