Learn More
Biomatik Corporation Mouse IGF1 (Insulin-like growth factor 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57760
Encompass_Preferred
Mouse IGF1 (Insulin-like growth factor 1) ELISA Kit for detecting IGF1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05017. Target Synonyms: IGF-1; IGF1; IGFI; IGF-I; IGF1A; IGF-IA; IGF-IB; MGF; Somatomedin C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0932