Learn More
Biomatik Corporation Mouse IL-1b (Interleukin 1 Beta) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57763
Encompass_Preferred
Mouse IL-1β (Interleukin 1 Beta) ELISA Kit for detecting IL-1β in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800pg/ml. Sensitivity: 7.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10749. Target Synonyms: IL-1β; Interleukin 1 Beta; IL-1beta; IL1B; IL1-BETA; IL1F2; catabolin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0935