Learn More
Biomatik Corporation Mouse IL-4 (Interleukin 4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57766
Encompass_Preferred
Mouse IL-4 (Interleukin 4) ELISA Kit for detecting IL-4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07750. Target Synonyms: IL-4; Interleukin 4; IL4; BCGF-1; BCGF1; BSF-1; BSF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0938