Learn More
Biomatik Corporation Mouse IL-6 (Interleukin 6) ELISA Kit, 96 tests
Catalog No. 501500939
Encompass_Preferred
Mouse IL-6 (Interleukin 6) ELISA Kit for detecting IL-6 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08505. Target Synonyms: IL-6; Interleukin 6; IL6; BSF2; HGF; HSF; IFNB2; CDF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0939 Supplier Biomatik Corporation Supplier No. EKF57767
Mouse IL-6 (Interleukin 6) ELISA Kit for detecting IL-6 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08505. Target Synonyms: IL-6; Interleukin 6; IL6; BSF2; HGF; HSF; IFNB2; CDF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.