Learn More
Biomatik Corporation Mouse Leptin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57769
Encompass_Preferred
Mouse Leptin ELISA Kit for detecting Leptin in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P41160. Target Synonyms: Leptin; LEP; LEPD; OBS; Obesity Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0941