Learn More
Biomatik Corporation Mouse TGF-1 (Transforming Growth Factor 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57778
Encompass_Preferred
Mouse TGF-β1 (Transforming Growth Factor β1) ELISA Kit for detecting TGF-β1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04202. Target Synonyms: TGF-β1; Transforming Growth Factor Beta 1; TGF-B1; TGFB; TGFbeta; CED; DPD1; TGFβ1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0950