Learn More
Biomatik Corporation Mouse TGF-3 (Transforming Growth Factor 3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57779
Encompass_Preferred
Mouse TGF-β3 (Transforming Growth Factor β3) ELISA Kit for detecting TGF-β3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P17125. Target Synonyms: TGF-β3; Transforming Growth Factor Beta 3; TGFβ3; TGFB3; ARVD; FLJ16571; TGF-beta3; TGF-beta-3; transforming growth factor beta-3; transforming growth factor; beta 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0951