Learn More
Biomatik Corporation Mouse TNF (Tumor Necrosis Factor Alpha) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57781
Encompass_Preferred
Mouse TNF-α (Tumor Necrosis Factor Alpha) ELISA Kit for detecting TNF-α in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.906-250pg/ml. Sensitivity: 2.344pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06804. Target Synonyms: TNFα; Tumor Necrosis Factor Alpha; TNF-α; DIF; TNF-alpha; TNFA; TNFSF2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0953