Learn More
Biomatik Corporation Mouse TPO (Thrombopoietin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57782
Encompass_Preferred
Mouse TPO (Thrombopoietin) ELISA Kit for detecting TPO in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.125-200pg/ml. Sensitivity: 1.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35419. Target Synonyms: TPO; Thrombopoietin; MGDF; MK-CSF; THPO. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0954