Learn More
Biomatik Corporation Mouse VEGF (Vascular Endothelial cell Growth Factor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57784
Encompass_Preferred
Mouse VEGF (Vascular Endothelial cell Growth Factor) ELISA Kit for detecting VEGF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q00731. Target Synonyms: VEGF; Vascular Endothelial Cell Growth Factor; VAS; Vasculotropin; VEGFA; VPF; MVCD1; vascular endothelial growth factor A; Vascular permeability factor; VEGF-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0956