Learn More
Biomatik Corporation Mouse Cga (Glycoprotein hormones alpha chain) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57790
Encompass_Preferred
Mouse Cga (Glycoprotein hormones alpha chain) ELISA Kit for detecting Cga in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.125-200uiu/ml. Sensitivity: 1.875uiu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01216. Target Synonyms: Cga; Glycoprotein hormones alpha chain; Glycoprotein hormones alpha chain; Thyroid-stimulating hormone alpha chain; TSH-alpha; Lutropin alpha chain; Luteinizing hormone alpha chain; LSH-alpha; Follitropin alpha chain; Thyrotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha; Follicle-stimulating hormone alpha chain; FSH-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0962