Learn More
Biomatik Corporation Mouse MTOR (Serine/threonine-protein kinase mTOR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57793
Encompass_Preferred
Mouse MTOR (Serine/threonine-protein kinase mTOR) ELISA Kit for detecting MTOR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9JLN9. Target Synonyms: Serine; threonine-protein kinase mTOR; Rapamycin target protein 1; Rapamycin target protein 1; RAPT1; Mechanistic target of rapamycin; Mammalian target of rapamycin; mTOR; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FRAP; FRAP1; FRAP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0965