Learn More
Biomatik Corporation Mouse Fgf21 (Fibroblast growth factor 21) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57794
Encompass_Preferred
Mouse Fgf21 (Fibroblast growth factor 21) ELISA Kit for detecting Fgf21 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9JJN1. Target Synonyms: FGF21; FGF-21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0966